siRNA Details
| Gene Symbol: VPS37C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017966.4 | NP_060436.4 | 5 | 2474 | ||
| XM_005274077.3 | XP_005274134.1 | 5 | 2515 | |||
| XM_017017955.1 | XP_016873444.1 | 2 | 2184 | |||
| GenBank mRNA | AK000854.1 | - | 1 | 1062 | ||
| AL834261.1 | - | 5 | 2482 | |||
| BC005805.2 | - | 1 | 1101 | |||
| BC062604.1 | - | 1 | 1197 | |||
| BC141827.1 | - | 5 | 2437 | 40148285 | ||
| BC142702.1 | - | 5 | 2437 | 40148798 | ||

VPS37C
Genomic Map