siRNA Details
| Gene Symbol: MGC108823 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012353.1 | NP_001012353.1 | 2 | 1474 | ||
| GenBank mRNA | BC089836.1 | - | 2 | 1474 | 7326302 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGC108823| Gene Symbol: MGC108823 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012353.1 | NP_001012353.1 | 2 | 1474 | ||
| GenBank mRNA | BC089836.1 | - | 2 | 1474 | 7326302 | |