siRNA Details
| Gene Symbol: NRDE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017970.3 | NP_060440.2 | 12 | 3519 | ||
| XM_011536886.1 | XP_011535188.1 | 13 | 3347 | |||
| XM_017021394.1 | XP_016876883.1 | 13 | 3347 | |||
| XM_017021395.1 | XP_016876884.1 | 13 | 3468 | |||
| XM_017021396.1 | XP_016876885.1 | 12 | 3062 | |||
| GenBank mRNA | AK024113.1 | - | 7 | 2060 | ||
| AK294958.1 | - | 12 | 3323 | |||
| AL832941.1 | - | 2 | 492 | |||
| BC098568.1 | - | 12 | 3297 | 30706812 | ||
| BC118673.1 | - | 9 | 2852 | 40028659 | ||

NRDE2
Genomic Map