siRNA Details
| Gene Symbol: Kctd14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010826.3 | NP_001010826.2 | 3 | 931 | ||
| NM_001012434.3 | NP_001012436.1 | 3 | 964 | |||
| NM_001136235.1 | NP_001129707.1 | 3 | 948 | |||
| GenBank mRNA | AK145279.1 | - | 3 | 932 | ||
| AY899809.1 | - | 3 | 964 | |||
| BC024588.1 | - | 3 | 803 | 5066294 | ||

Kctd14
Genomic Map