siRNA Details
| Gene Symbol: Rnase10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012467.1 | NP_001012485.1 | 1 | 316 | ||
| XM_006251868.2 | XP_006251930.1 | 2 | 1823 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnase10| Gene Symbol: Rnase10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012467.1 | NP_001012485.1 | 1 | 316 | ||
| XM_006251868.2 | XP_006251930.1 | 2 | 1823 | |||