siRNA Details
| Gene Symbol: Wee1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012742.1 | NP_001012760.1 | 11 | 2075 | ||
| GenBank mRNA | BC090346.1 | - | 11 | 2075 | 7306767 | |
| D31838.1 | - | 11 | 2274 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wee1| Gene Symbol: Wee1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012742.1 | NP_001012760.1 | 11 | 2075 | ||
| GenBank mRNA | BC090346.1 | - | 11 | 2075 | 7306767 | |
| D31838.1 | - | 11 | 2274 | |||