siRNA Details
| Gene Symbol: Pip5k1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012743.1 | NP_001012761.1 | 15 | 2626 | ||
| GenBank mRNA | BC090349.1 | - | 15 | 2626 | 7311399 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pip5k1b| Gene Symbol: Pip5k1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012743.1 | NP_001012761.1 | 15 | 2626 | ||
| GenBank mRNA | BC090349.1 | - | 15 | 2626 | 7311399 | |