siRNA Details
| Gene Symbol: Gmpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013036.1 | NP_001013054.1 | 10 | 986 | ||
| GenBank mRNA | BC089848.1 | - | 10 | 986 | 7374684 | |
| FQ211507.1 | - | 11 | 1898 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gmpr2| Gene Symbol: Gmpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013036.1 | NP_001013054.1 | 10 | 986 | ||
| GenBank mRNA | BC089848.1 | - | 10 | 986 | 7374684 | |
| FQ211507.1 | - | 11 | 1898 | |||