siRNA Details
| Gene Symbol: Allc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013037.1 | NP_001013055.1 | 3, 4 | 304 | ||
| XM_006239944.2 | XP_006240006.1 | 3, 4 | 378 | |||
| GenBank mRNA | BC078971.1 | - | 3, 4 | 304 | 7111813 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Allc| Gene Symbol: Allc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013037.1 | NP_001013055.1 | 3, 4 | 304 | ||
| XM_006239944.2 | XP_006240006.1 | 3, 4 | 378 | |||
| GenBank mRNA | BC078971.1 | - | 3, 4 | 304 | 7111813 | |