siRNA Details
Gene Symbol: Gdf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013038.1 | NP_001013056.1 | 1 | 427 | ||
GenBank mRNA | AJ537426.2 | - | 1 | 427 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gdf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013038.1 | NP_001013056.1 | 1 | 427 | ||
GenBank mRNA | AJ537426.2 | - | 1 | 427 |