siRNA Details
| Gene Symbol: Galt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013089.2 | NP_001013107.1 | 3 | 355 | ||
| XM_006238033.2 | XP_006238095.1 | 3 | 279 | |||
| XM_008763615.1 | XP_008761837.1 | 2 | 124 | |||
| GenBank mRNA | BC098756.1 | - | 3 | 355 | 7370278 | |

Galt
Genomic Map