siRNA Details
| Gene Symbol: Galt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013089.2 | NP_001013107.1 | 2 | 194 | ||
| XM_006238033.2 | XP_006238095.1 | 2 | 118 | |||
| GenBank mRNA | BC098756.1 | - | 2 | 194 | 7370278 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Galt| Gene Symbol: Galt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013089.2 | NP_001013107.1 | 2 | 194 | ||
| XM_006238033.2 | XP_006238095.1 | 2 | 118 | |||
| GenBank mRNA | BC098756.1 | - | 2 | 194 | 7370278 | |