siRNA Details
| Gene Symbol: Trmt10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013090.2 | NP_001013108.1 | 8 | 958 | ||
| XM_006238042.1 | XP_006238104.1 | 9 | 1008 | |||
| XM_006238043.1 | XP_006238105.1 | 9 | 1207 | |||
| XM_006238044.2 | XP_006238106.1 | 9 | 968 | |||
| GenBank mRNA | BC086603.1 | - | 8 | 962 | 7315452 | |

Trmt10b
Genomic Map