siRNA Details
| Gene Symbol: Eif3g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013095.1 | NP_001013113.1 | 10 | 936 | ||
| GenBank mRNA | BC086383.1 | - | 10 | 936 | 7301572 | |
| FQ213005.1 | - | 10 | 977 | |||
| FQ225883.1 | - | 10 | 969 | |||
| FQ226276.1 | - | 10 | 968 | |||
| FQ229703.1 | - | 10 | 967 | |||
| FQ230888.1 | - | 10 | 962 | |||
| FQ232595.1 | - | 10 | 966 | |||
| FQ232661.1 | - | 10 | 966 | |||
| FQ234018.1 | - | 10 | 970 | |||
| FQ234727.1 | - | 8 | 818 | |||
| FQ234921.1 | - | 10 | 967 | |||
| FQ234929.1 | - | 10 | 969 | |||
| FQ235264.1 | - | 7 | 750 | |||

Eif3g
Genomic Map