siRNA Details
| Gene Symbol: Mycn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013096.1 | NP_001013114.1 | 3 | 2375 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mycn| Gene Symbol: Mycn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013096.1 | NP_001013114.1 | 3 | 2375 | ||