siRNA Details
| Gene Symbol: Snapc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013121.1 | NP_001013139.1 | 5 | 800 | ||
| XM_008768956.1 | XP_008767178.1 | 5 | 903 | |||
| GenBank mRNA | BC079077.1 | - | 5 | 800 | 7122319 | |
| FQ214278.1 | - | 4 | 630 | |||

Snapc2
Genomic Map