siRNA Details
| Gene Symbol: Snapc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013121.1 | NP_001013139.1 | 1 | 119 | ||
| XM_008768956.1 | XP_008767178.1 | 1 | 171 | |||
| GenBank mRNA | BC079077.1 | - | 1 | 119 | 7122319 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snapc2| Gene Symbol: Snapc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013121.1 | NP_001013139.1 | 1 | 119 | ||
| XM_008768956.1 | XP_008767178.1 | 1 | 171 | |||
| GenBank mRNA | BC079077.1 | - | 1 | 119 | 7122319 | |