siRNA Details
| Gene Symbol: Dusp18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013128.2 | NP_001013146.2 | 3 | 804 | ||
| XM_006251299.2 | XP_006251361.1 | 3 | 1228 | |||
| GenBank mRNA | BC079285.1 | - | 3 | 786 | 7134264 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dusp18| Gene Symbol: Dusp18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013128.2 | NP_001013146.2 | 3 | 804 | ||
| XM_006251299.2 | XP_006251361.1 | 3 | 1228 | |||
| GenBank mRNA | BC079285.1 | - | 3 | 786 | 7134264 | |