siRNA Details
| Gene Symbol: Fars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013139.1 | NP_001013157.1 | 8 | 1386 | ||
| XM_006253823.2 | XP_006253885.1 | 8 | 1507 | |||
| XM_006253825.2 | XP_006253887.1 | 7 | 1444 | |||
| XM_008771572.1 | XP_008769794.1 | 9 | 1694 | |||
| XM_008771573.1 | XP_008769795.1 | 9 | 2370 | |||
| XM_008771574.1 | XP_008769796.1 | 8 | 1429 | |||
| XM_008771575.1 | XP_008769797.1 | 9 | 1636 | |||
| XM_008771576.1 | XP_008769798.1 | 9 | 1593 | |||
| XM_008771577.1 | XP_008769799.1 | 7 | 1444 | |||
| GenBank mRNA | BC078956.1 | - | 8 | 1386 | 7110464 | |
| FQ232145.1 | - | 7 | 1363 | |||

Fars2
Genomic Map