siRNA Details
| Gene Symbol: Zfp35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013141.1 | NP_001013159.1 | 3 | 1009 | ||
| GenBank mRNA | BC089850.1 | - | 3 | 1009 | 7375328 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp35| Gene Symbol: Zfp35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013141.1 | NP_001013159.1 | 3 | 1009 | ||
| GenBank mRNA | BC089850.1 | - | 3 | 1009 | 7375328 | |