siRNA Details
| Gene Symbol: Plekhf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013148.1 | NP_001013166.1 | 1 | 406 | ||
| XM_006228905.2 | XP_006228967.1 | 2 | 501 | |||
| GenBank mRNA | BC079354.1 | - | 1 | 406 | 7108247 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plekhf1| Gene Symbol: Plekhf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013148.1 | NP_001013166.1 | 1 | 406 | ||
| XM_006228905.2 | XP_006228967.1 | 2 | 501 | |||
| GenBank mRNA | BC079354.1 | - | 1 | 406 | 7108247 | |