siRNA Details
| Gene Symbol: MREG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018000.2 | NP_060470.2 | 5 | 2587 | ||
| XM_011511468.2 | XP_011509770.1 | 6 | 2587 | |||
| GenBank mRNA | AK000978.1 | - | 6 | 1122 | ||
| BC032747.1 | - | 6 | 1511 | 5573149 | ||
| BC082990.1 | - | 6 | 2206 | 6159624 | ||

MREG
Genomic Map