siRNA Details
| Gene Symbol: Scrn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013162.1 | NP_001013180.1 | 4 | 506 | ||
| XM_006234358.2 | XP_006234420.1 | 4 | 543 | |||
| XM_006234359.2 | XP_006234421.1 | 4 | 545 | |||
| XM_006234360.2 | XP_006234422.1 | 4 | 545 | |||
| GenBank mRNA | BC088234.1 | - | 4 | 506 | 7324882 | |

Scrn3
Genomic Map