siRNA Details
| Gene Symbol: Scrn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013162.1 | NP_001013180.1 | 6 | 968 | ||
| GenBank mRNA | BC088234.1 | - | 6 | 968 | 7324882 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scrn3| Gene Symbol: Scrn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013162.1 | NP_001013180.1 | 6 | 968 | ||
| GenBank mRNA | BC088234.1 | - | 6 | 968 | 7324882 | |