siRNA Details
| Gene Symbol: Wasf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013167.1 | NP_001013185.1 | 7 | 1338 | ||
| GenBank mRNA | BC089207.1 | - | 7 | 1338 | 7135528 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wasf2| Gene Symbol: Wasf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013167.1 | NP_001013185.1 | 7 | 1338 | ||
| GenBank mRNA | BC089207.1 | - | 7 | 1338 | 7135528 | |