siRNA Details
| Gene Symbol: Wars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013170.2 | NP_001013188.2 | 6 | 775 | ||
| XM_006240542.2 | XP_006240604.1 | 6 | 812 | |||
| XM_008764811.1 | XP_008763033.1 | 6 | 966 | |||
| XM_008764812.1 | XP_008763034.1 | 6 | 1563 | |||
| GenBank mRNA | AY724504.1 | - | 6 | 775 | ||
| BC061752.1 | - | 7 | 838 | 5600013 | ||

Wars
Genomic Map