siRNA Details
Gene Symbol: Btg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013176.1 | NP_001013194.1 | 3, 4 | 503 | ||
GenBank mRNA | BC088755.1 | - | 3, 4 | 503 | 7307514 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Btg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013176.1 | NP_001013194.1 | 3, 4 | 503 | ||
GenBank mRNA | BC088755.1 | - | 3, 4 | 503 | 7307514 |