siRNA Details
| Gene Symbol: Sult1c2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013177.2 | NP_001013195.2 | 2710 | |||
| GenBank mRNA | BC092564.1 | - | 2710 | 7372924 | ||
| FQ209913.1 | - | 2704 | ||||
| Gene Symbol: RGD1559960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006244256.2 | XP_006244318.2 | 5 | 2395 | ||
| XM_008766792.1 | XP_008765014.1 | 6 | 2542 | |||

Sult1c2a
Genomic Map