siRNA Details
| Gene Symbol: Zbtb16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013181.1 | NP_001013199.1 | 2 | 237 | ||
| GenBank mRNA | AY781102.1 | - | 2 | 237 | ||
| AY781103.1 | - | 2 | 237 | |||
| AY781104.1 | - | 2 | 228 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb16| Gene Symbol: Zbtb16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013181.1 | NP_001013199.1 | 2 | 237 | ||
| GenBank mRNA | AY781102.1 | - | 2 | 237 | ||
| AY781103.1 | - | 2 | 237 | |||
| AY781104.1 | - | 2 | 228 | |||