siRNA Details
| Gene Symbol: Narfl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013183.1 | NP_001013201.1 | 4 | 412 | ||
| GenBank mRNA | BC091240.1 | - | 4 | 412 | 7325087 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Narfl| Gene Symbol: Narfl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013183.1 | NP_001013201.1 | 4 | 412 | ||
| GenBank mRNA | BC091240.1 | - | 4 | 412 | 7325087 | |