siRNA Details
| Gene Symbol: IFT57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018010.3 | NP_060480.1 | 3 | 671 | ||
| GenBank mRNA | AB014762.1 | - | 3 | 423 | ||
| AF139576.1 | - | 3 | 501 | |||
| AF245220.1 | - | 3 | 565 | |||
| AK001009.1 | - | 3 | 510 | |||
| AK298854.1 | - | 3 | 569 | |||
| BC011899.2 | - | 3 | 520 | 4053749 | ||
| CR457251.1 | - | 3 | 423 | |||

IFT57
Genomic Map