siRNA Details
| Gene Symbol: Podnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013384.2 | NP_001013402.2 | 2 | 294 | ||
| GenBank mRNA | BC063764.1 | - | 2 | 294 | 30288186 | |
| DQ303463.1 | - | 2 | 279 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Podnl1| Gene Symbol: Podnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013384.2 | NP_001013402.2 | 2 | 294 | ||
| GenBank mRNA | BC063764.1 | - | 2 | 294 | 30288186 | |
| DQ303463.1 | - | 2 | 279 | |||