siRNA Details
| Gene Symbol: Rhoh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013430.1 | NP_001013448.1 | 3 | 1473 | ||
| GenBank mRNA | BC091269.1 | - | 3 | 1473 | 7381512 | |
| FQ225173.1 | - | 3 | 1485 | |||
| FQ232983.1 | - | 3 | 1485 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhoh| Gene Symbol: Rhoh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013430.1 | NP_001013448.1 | 3 | 1473 | ||
| GenBank mRNA | BC091269.1 | - | 3 | 1473 | 7381512 | |
| FQ225173.1 | - | 3 | 1485 | |||
| FQ232983.1 | - | 3 | 1485 | |||