siRNA Details
| Gene Symbol: Chchd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013431.1 | NP_001013449.1 | 3 | 900 | ||
| GenBank mRNA | BC091407.1 | - | 3 | 900 | 7324948 | |
| FQ220478.1 | - | 3 | 765 | |||
| FQ220795.1 | - | 3 | 795 | |||
| FQ224864.1 | - | 3 | 814 | |||
| FQ229282.1 | - | 3 | 765 | |||

Chchd4
Genomic Map