siRNA Details
| Gene Symbol: EPGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005265666.4 | XP_005265723.1 | 4 | 1571 | ||
| GenBank mRNA | AY358920.1 | - | 4 | 655 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EPGN| Gene Symbol: EPGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005265666.4 | XP_005265723.1 | 4 | 1571 | ||
| GenBank mRNA | AY358920.1 | - | 4 | 655 | ||