siRNA Details
| Gene Symbol: CHIAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003928.2 | 8 | 2514 | |||
| GenBank mRNA | BC031662.2 | - | 8 | 2514 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHIAP2| Gene Symbol: CHIAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003928.2 | 8 | 2514 | |||
| GenBank mRNA | BC031662.2 | - | 8 | 2514 | ||