siRNA Details
| Gene Symbol: CDK17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170464.2 | NP_001163935.1 | 4 | 1013 | ||
| NM_002595.4 | NP_002586.2 | 4 | 1013 | |||
| XM_017019405.1 | XP_016874894.1 | 4 | 455 | |||
| XM_017019406.1 | XP_016874895.1 | 4 | 470 | |||
| XM_017019407.1 | XP_016874896.1 | 4 | 900 | |||
| GenBank mRNA | AK290011.1 | - | 4 | 905 | ||
| AK294698.1 | - | 3 | 352 | |||
| AK315214.1 | - | 4 | 923 | |||
| BC033005.1 | - | 4 | 502 | 4824359 | ||
| X66360.1 | - | 4 | 450 | |||

CDK17
Genomic Map