siRNA Details
Gene Symbol: LINC00268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126863.1 | - | 1 | 542 | ||
BC136786.1 | - | 1 | 424 | 9020776 | ||
BC136790.1 | - | 1 | 425 | 9020780 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126863.1 | - | 1 | 542 | ||
BC136786.1 | - | 1 | 424 | 9020776 | ||
BC136790.1 | - | 1 | 425 | 9020780 |