siRNA Details
| Gene Symbol: DGKK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013742.3 | NP_001013764.1 | 9 | 1555 | ||
| XM_017029268.1 | XP_016884757.1 | 9 | 1667 | |||
| GenBank mRNA | AB183864.1 | - | 9 | 1555 | ||
| BC137319.1 | - | 9 | 1539 | 9021316 | ||
| BC137320.1 | - | 9 | 1539 | 9021317 | ||

DGKK
Genomic Map