siRNA Details
| Gene Symbol: Gas2l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013759.2 | NP_001013781.1 | 6 | 2666 | ||
| GenBank mRNA | BB806501.1 | - | 1 | 108 | ||
| BC147480.1 | - | 6 | 2666 | 9087436 | ||
| BC150847.1 | - | 6 | 2666 | 9087758 | ||

Gas2l2
Genomic Map