siRNA Details
| Gene Symbol: Cldn34b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001097980.1 | NP_001091449.1 | 2 | 364 | ||
| Gene Symbol: Cldn34b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081671.1 | NP_001075140.1 | 3 | 408 | ||
| GenBank mRNA | AK006680.1 | - | 2 | 366 | ||
| BC100353.1 | - | 3 | 408 | 30933439 | ||
| Gene Symbol: Cldn34b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013760.2 | NP_001013782.1 | 2 | 366 | ||
| GenBank mRNA | BC048515.1 | - | 2 | 366 | 6704957 | |

Cldn34b1
Genomic Map