siRNA Details
| Gene Symbol: Fggy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013932.2 | NP_001013954.2 | 16 | 1715 | ||
| XM_006238410.2 | XP_006238472.1 | 16 | 2353 | |||
| XM_006238413.2 | XP_006238475.1 | 17 | 1926 | |||
| XM_008763856.1 | XP_008762078.1 | 16 | 1715 | |||
| XM_008763857.1 | XP_008762079.1 | 11 | 1817 | |||
| XM_008763858.1 | XP_008762080.1 | 12 | 3107 | |||
| XM_008763859.1 | XP_008762081.1 | 12 | 1564 | |||
| GenBank mRNA | BC090077.1 | - | 15 | 1540 | 7107754 | |

Fggy
Genomic Map