siRNA Details
| Gene Symbol: Dync2li1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013940.1 | NP_001013962.1 | 11, 12 | 1378 | ||
| XM_006239695.2 | XP_006239757.1 | 12, 13 | 1138 | |||
| GenBank mRNA | BC079201.1 | - | 11, 12 | 1378 | 7131728 | |

Dync2li1
Genomic Map