siRNA Details
| Gene Symbol: LOC298795 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013941.1 | NP_001013963.1 | 1 | 1251 | ||
| GenBank mRNA | BC079389.1 | - | 1 | 1240 | ||
| BC089860.1 | - | 1 | 1251 | 7112967 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC298795| Gene Symbol: LOC298795 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013941.1 | NP_001013963.1 | 1 | 1251 | ||
| GenBank mRNA | BC079389.1 | - | 1 | 1240 | ||
| BC089860.1 | - | 1 | 1251 | 7112967 | ||