siRNA Details
Gene Symbol: LOC298795 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013941.1 | NP_001013963.1 | 1 | 1228 | ||
GenBank mRNA | BC079389.1 | - | 1 | 1217 | ||
BC089860.1 | - | 1 | 1228 | 7112967 |
Gene Symbol: Sfn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750048.3 | XP_003750096.1 | 1 | 1465 |
