siRNA Details
| Gene Symbol: Map3k7cl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013979.1 | NP_001014001.1 | 5 | 1149 | ||
| XM_006248016.2 | XP_006248078.1 | 6 | 1861 | |||
| GenBank mRNA | BC076389.1 | - | 5 | 1149 | 7110014 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map3k7cl| Gene Symbol: Map3k7cl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013979.1 | NP_001014001.1 | 5 | 1149 | ||
| XM_006248016.2 | XP_006248078.1 | 6 | 1861 | |||
| GenBank mRNA | BC076389.1 | - | 5 | 1149 | 7110014 | |