siRNA Details
| Gene Symbol: Cyyr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013980.1 | NP_001014002.1 | 4 | 3003 | ||
| GenBank mRNA | BC087052.1 | - | 4 | 3003 | 7122930 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyyr1| Gene Symbol: Cyyr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013980.1 | NP_001014002.1 | 4 | 3003 | ||
| GenBank mRNA | BC087052.1 | - | 4 | 3003 | 7122930 | |