siRNA Details
| Gene Symbol: Tmem175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013991.1 | NP_001014013.1 | 11 | 1664 | ||
| XM_006250558.2 | XP_006250620.1 | 11 | 2120 | |||
| XM_008769936.1 | XP_008768158.1 | 9 | 2347 | |||
| XM_008769937.1 | XP_008768159.1 | 8 | 1585 | |||
| GenBank mRNA | BC079245.1 | - | 11 | 1664 | 7132812 | |

Tmem175
Genomic Map