siRNA Details
| Gene Symbol: LOC305806 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013993.1 | NP_001014015.1 | 889 | |||
| GenBank mRNA | BC079250.1 | - | 889 | 7133060 | ||
| Gene Symbol: LOC290508 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC079266.1 | - | 927 | 7133637 | ||

LOC305806
Genomic Map