siRNA Details
| Gene Symbol: Slc25a37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013996.1 | NP_001014018.1 | 3, 4 | 638 | ||
| XM_006252266.1 | XP_006252328.1 | 4, 5 | 810 | |||
| GenBank mRNA | BC082071.1 | - | 3, 4 | 638 | 7132913 | |
| FQ227341.1 | - | 3, 4 | 579 | |||
| FQ230701.1 | - | 2, 3 | 295 | |||
| FQ231671.1 | - | 2, 3 | 109 | |||
| FQ231945.1 | - | 2, 3 | 126 | |||
| FQ233331.1 | - | 3, 4 | 538 | |||

Slc25a37
Genomic Map